Gene Transcripts

Gene Symbol
Ndufa12
Entrez Gene ID
66414
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12
Gene Aliases
2410011G03Rik, AW112974
Chromosome Location
Chr. 10: 94199009 - 94220948 on Build GRCm38
UniGene ID
Mm.27886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025551.3 NP_079827.2 1, 2 130
GenBank mRNA AK003131.1 - 1, 2 103
AK003322.1 - 1, 2 103
AK008876.1 - 1, 2 103
AK010460.1 - 1, 2 131
AK032774.1 - 1, 2 104
AK134765.1 - 1, 2 103
AK153240.1 - 1, 2 103
AK167943.1 - 1, 2 96
BC004633.1 - 1560
BC086922.1 - 1, 2 135 6751147

View Full Product Details