Gene Transcripts
- Gene Symbol
- Lrrc57
- Entrez Gene ID
- 66606
- Gene Name
- leucine rich repeat containing 57
- Gene Aliases
- 2810002D13Rik, AA407405
- Chromosome Location
- Chr. 2: 120604238 - 120609508 on Build GRCm38
- UniGene ID
- Mm.428639
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159609.1 |
NP_001153081.1
|
5, 6 |
873 |
|
NM_001159610.1 |
NP_001153082.1
|
5, 6 |
924 |
|
NM_001159612.1 |
NP_001153084.1
|
5, 6 |
940 |
|
NM_025657.3 |
NP_079933.2
|
5, 6 |
823 |
|
GenBank mRNA |
AK003037.1 |
- |
5, 6 |
730 |
|
AK003592.1 |
- |
5, 6 |
841 |
|
AK012637.1 |
- |
5, 6 |
937 |
|
AK015684.1 |
- |
5, 6 |
875 |
|
AK134971.1 |
- |
5, 6 |
782 |
|
AK169249.1 |
- |
5, 6 |
847 |
|
BC034894.1 |
- |
5, 6 |
782 |
3328402 |
View Full Product Details