Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025928.3 | NP_080204.1 | 2 | 178 | |
GenBank mRNA | AF348510.1 | - | 2 | 188 | |
AK011174.1 | - | 2 | 180 | ||
AK012113.1 | - | 2 | 179 | ||
AK013787.1 | - | 2 | 181 | ||
AK078308.1 | - | 2 | 197 | ||
AK150603.1 | - | 2 | 195 | ||
AK168200.1 | - | 2 | 179 | ||
BC034333.1 | - | 2 | 156 | ||
BC037139.1 | - | 2 | 187 | 4952550 | |
BC049223.1 | - | 2 | 177 | 3257320 |