Gene Transcripts

Gene Symbol
Mto1
Entrez Gene ID
68291
Gene Name
mitochondrial translation optimization 1 homolog (S. cerevisiae)
Gene Aliases
2310039H01Rik, 5730419A02Rik
Chromosome Location
Chr. 9: 78448015 - 78474152 on Build GRCm38
UniGene ID
Mm.291779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026658.2 NP_080934.2 6 1166
GenBank mRNA AF369902.1 - 6 1156
AK019088.1 - 6 1166
BC063256.1 - 6 1152

View Full Product Details