Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027460.2 | NP_081736.2 | 7 | 1415 | |
GenBank mRNA | AK017628.1 | - | 7 | 1326 | |
AK157423.1 | - | 7 | 1416 | ||
BB772382.1 | - | 2 | 512 | ||
BC011293.1 | - | 7 | 1390 | 3989313 |