Gene Transcripts

Gene Symbol
MBL2
Entrez Gene ID
4153
Gene Name
mannose binding lectin 2
Gene Aliases
COLEC1, HSMBPC, MBL, MBL2D, MBP, MBP-C, MBP1, MBPD
Chromosome Location
Chr. 10: 52764977 - 52772845 on Build GRCh38
UniGene ID
Hs.499674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000242.2 NP_000233.1 4 3430
XM_006717861.3 XP_006717924.1 5 3498
XM_011539816.2 XP_011538118.1 5 3482
GenBank mRNA X15422.1 - 4 3430

View Full Product Details