Gene Transcripts

Gene Symbol
Tsga13
Entrez Gene ID
116732
Gene Name
testis specific gene A13
Gene Aliases
1700023D02Rik, Vme1
Chromosome Location
Chr. 6: 30896980 - 30925154 on Build GRCm38
UniGene ID
Mm.207048
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054073.2 NP_473414.1 5 495
GenBank mRNA AF294348.1 - 5 463
AK006265.1 - 5 496
BC125597.1 - 4 413 40130112
BC125599.1 - 4 413 40130114

View Full Product Details