Gene Transcripts

Gene Symbol
Miip
Entrez Gene ID
28010
Gene Name
migration and invasion inhibitory protein
Gene Aliases
AA553001, AI853621, D4Wsu114e, IIP45
Chromosome Location
Chr. 4: 147860778 - 147868719 on Build GRCm38
UniGene ID
Mm.250504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC017525.1 - 1871
BC024069.1 - 1581 5097088
FJ618908.1 - 1799

View Full Product Details