Gene Transcripts
- Gene Symbol
- Oas1a
- Entrez Gene ID
- 246730
- Gene Name
- 2'-5' oligoadenylate synthetase 1A
- Gene Aliases
- L3
- Chromosome Location
- Chr. 5: 120896256 - 120907543 on Build GRCm38
- UniGene ID
- Mm.14301
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC013715.1 |
- |
7 |
1660 |
|
View Full Product Details