Gene Transcripts

Gene Symbol
Mcmbp
Entrez Gene ID
210711
Gene Name
MCM (minichromosome maintenance deficient) binding protein
Gene Aliases
1110007A13Rik, AU017382, BC025641, MCM-BP
Chromosome Location
Chr. 7: 128696441 - 128740429 on Build GRCm38
UniGene ID
Mm.490344
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145955.3 NP_666067.1 16 3480
GenBank mRNA AK142281.1 - 1 455
AK169563.1 - 464
BC038342.1 - 16 3480 4191750

View Full Product Details