Gene Transcripts

Gene Symbol
Rdh19
Entrez Gene ID
216453
Gene Name
retinol dehydrogenase 19
Gene Aliases
RDH-S, RDH-S2, Rdhs
Chromosome Location
Chr. 10: 127849928 - 127861176 on Build GRCm38
UniGene ID
Mm.222953
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147222.2 NP_671755.2 4 1381
GenBank mRNA AY046408.1 - 4 1378

View Full Product Details