Gene Transcripts
- Gene Symbol
- RPL3
- Entrez Gene ID
- 6122
- Gene Name
- ribosomal protein L3
- Gene Aliases
- ASC-1, L3, TARBP-B
- Chromosome Location
- Chr. 22: 39312882 - 39319665 on Build GRCh38
- UniGene ID
- Hs.119598
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000967.3 |
NP_000958.1
|
2, 3 |
274 |
|
NM_001033853.1 |
NP_001029025.1
|
2, 3 |
274 |
|
GenBank mRNA |
AB062291.1 |
- |
1, 2 |
211 |
|
AB266619.1 |
- |
1, 2 |
50 |
|
AK092695.1 |
- |
2, 3 |
232 |
|
AK297711.1 |
- |
2, 3 |
232 |
|
AK315693.1 |
- |
2, 3 |
232 |
|
BC002408.2 |
- |
1, 2 |
209 |
2961541 |
BC006483.1 |
- |
2, 3 |
219 |
|
BC008003.1 |
- |
1, 2 |
211 |
3507481 |
BC008492.1 |
- |
2, 3 |
232 |
4251511 |
BC012146.1 |
- |
2, 3 |
219 |
4549682 |
BC012786.2 |
- |
2, 3 |
219 |
3951917 |
BC013674.1 |
- |
1, 2 |
215 |
4152521 |
BC013861.1 |
- |
1, 2 |
215 |
|
BC014017.2 |
- |
1, 2 |
214 |
4128023 |
BC015032.1 |
- |
1, 2 |
215 |
|
BC015767.1 |
- |
1, 2 |
215 |
4861546 |
BC022790.1 |
- |
1, 2 |
120 |
|
BC033296.1 |
- |
1, 2 |
212 |
|
BC036582.1 |
- |
2, 3 |
871 |
4796724 |
BC063662.1 |
- |
1, 2 |
219 |
3852576 |
BC088373.1 |
- |
2, 3 |
229 |
6195715 |
BC107711.1 |
- |
2, 3 |
236 |
6699816 |
CR456566.1 |
- |
1, 2 |
218 |
|
L22453.1 |
- |
1, 2 |
209 |
|
M90054.1 |
- |
1, 2 |
192 |
|
X73460.1 |
- |
1, 2 |
212 |
|
View Full Product Details