Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153136.4 | NP_694776.2 | 2, 3 | 486 | |
GenBank mRNA | AK089446.1 | - | 1, 2 | 274 | |
AK145997.1 | - | 2, 3 | 475 | ||
AK155086.1 | - | 2, 3 | 487 | ||
AK157937.1 | - | 2, 3 | 487 | ||
BC036718.1 | - | 2, 3 | 642 | 5322150 |