Gene Transcripts

Gene Symbol
Mmrn2
Entrez Gene ID
105450
Gene Name
multimerin 2
Gene Aliases
AA986839, ENDOGLYX1, Emilin3, EndoGlyx-1
Chromosome Location
Chr. 14: 34375504 - 34404287 on Build GRCm38
UniGene ID
Mm.272673
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153127.3 NP_694767.3 6 2444
GenBank mRNA AK077884.1 - 6 2445
AK078470.1 - 6 2420
AK134285.1 - 6 2483
BC034871.1 - 6 808
BC145845.1 - 6 2364 40131148

View Full Product Details