Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153388.4 | NP_700437.2 | 2 | 1167 | |
GenBank mRNA | AK081560.1 | - | 1 | 1450 | |
AK163603.1 | - | 2 | 1025 | ||
BC023036.1 | - | 1 | 780 | 5358628 | |
BC023156.1 | - | 2 | 823 | 4951386 | |
DQ078786.1 | - | 1 | 452 |