Gene Transcripts
- Gene Symbol
- Ubap2l
- Entrez Gene ID
- 74383
- Gene Name
- ubiquitin-associated protein 2-like
- Gene Aliases
- 3110083O19Rik, 4932431F02Rik, A430103N23Rik, C77168, Nice-4, mKIAA0144
- Chromosome Location
- Chr. 3: 89999589 - 90052623 on Build GRCm38
- UniGene ID
- Mm.25610
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165983.1 |
NP_001159455.1
|
3, 4 |
287 |
|
NM_001165984.1 |
NP_001159456.1
|
3, 4 |
262 |
|
NM_001165985.1 |
NP_001159457.1
|
3, 4 |
287 |
|
NM_001165986.1 |
NP_001159458.1
|
3, 4 |
262 |
|
NM_001165987.1 |
NP_001159459.1
|
3, 4 |
287 |
|
NM_001165988.1 |
NP_001159460.1
|
3, 4 |
262 |
|
NM_028475.2 |
NP_082751.1
|
3, 4 |
287 |
|
NM_153489.2 |
NP_705693.2
|
3, 4 |
287 |
|
GenBank mRNA |
AF276967.1 |
- |
3, 4 |
222 |
|
AK014274.1 |
- |
3, 4 |
224 |
|
AK030218.1 |
- |
3, 4 |
228 |
|
AK031934.1 |
- |
3, 4 |
267 |
|
AK040551.1 |
- |
3, 4 |
249 |
|
AK048286.1 |
- |
3, 4 |
288 |
|
AK079895.1 |
- |
3, 4 |
223 |
|
AK152135.1 |
- |
3, 4 |
241 |
|
AK190891.1 |
- |
1, 2 |
80 |
|
AK208584.1 |
- |
1, 2 |
80 |
|
BB648932.1 |
- |
3, 4 |
288 |
|
BB842333.1 |
- |
3, 4 |
262 |
|
BC023906.1 |
- |
3, 4 |
278 |
5321969 |
BC029075.1 |
- |
3, 4 |
277 |
4527901 |
BC050910.1 |
- |
2, 3 |
209 |
6417527 |
View Full Product Details