Gene Transcripts

Gene Symbol
Nhlrc3
Entrez Gene ID
212114
Gene Name
NHL repeat containing 3
Gene Aliases
4833441N19, 8030451K01Rik, mKIAA4083
Chromosome Location
Chr. 3: 53451996 - 53463258 on Build GRCm38
UniGene ID
Mm.85744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172501.2 NP_766089.1 7 1407
GenBank mRNA AK033167.1 - 7 1407
AK133986.1 - 1357
AK220477.1 - 1324
BC087903.1 - 7 1394 30250042

View Full Product Details