Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172715.3 | NP_766303.1 | 4 | 560 | |
GenBank mRNA | AK030171.1 | - | 5 | 745 | |
AK138410.1 | - | 3 | 720 | ||
AK160261.1 | - | 4 | 505 | ||
BC096769.1 | - | 4 | 493 | 30101305 | |
BC138228.1 | - | 3 | 313 | 8861247 | |
BC145669.1 | - | 3 | 313 | 40130972 |