Gene Transcripts

Gene Symbol
Strbp
Entrez Gene ID
20744
Gene Name
spermatid perinuclear RNA binding protein
Gene Aliases
6430510M02Rik, AI852513, C230082I21Rik, C86322, Spnr
Chromosome Location
Chr. 2: 37569868 - 37703969 on Build GRCm38
UniGene ID
Mm.237095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009261.3 NP_033287.2 17 8395
GenBank mRNA AK032246.1 - 1 2019
AK045193.1 - 1 1304
AY380798.1 - 1 2021

View Full Product Details