Gene Transcripts

Gene Symbol
H2-M10.5
Entrez Gene ID
224761
Gene Name
histocompatibility 2, M region locus 10.5
Gene Aliases
6.9H
Chromosome Location
Chr. 17: 36772910 - 36776235 on Build GRCm38
UniGene ID
Mm.246584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177637.3 NP_808305.2 3 577
GenBank mRNA AF539601.1 - 3 577
AY211082.1 - 3 586

View Full Product Details