Gene Transcripts

Gene Symbol
Ndor1
Entrez Gene ID
78797
Gene Name
NADPH dependent diflavin oxidoreductase 1
Gene Aliases
4930447P04Rik, NR1, Ndor
Chromosome Location
Chr. 2: 25244813 - 25255414 on Build GRCm38
UniGene ID
Mm.40020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK169885.1 - 2 265
AK211498.1 - 2 124
BC049789.1 - 2 256 6331033

View Full Product Details