Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005347.4 | NP_005338.1 | 5 | 1231 | |
GenBank mRNA | AF188611.1 | - | 5 | 916 | |
AF216292.1 | - | 5 | 970 | ||
AJ271729.1 | - | 5 | 983 | ||
AK129617.1 | - | 1 | 15 | ||
AK293604.1 | - | 6 | 1057 | ||
BC020235.1 | - | 5 | 1174 | 5020098 | |
EU794617.1 | - | 5 | 983 | ||
X87949.1 | - | 5 | 1189 |