Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204171.1 | NP_001191100.1 | 10 | 1256 | |
NM_001204172.1 | NP_001191101.1 | 3 | 428 | ||
NM_001278516.1 | NP_001265445.1 | 10 | 1338 | ||
NM_001278517.1 | NP_001265446.1 | 8 | 1112 | ||
NM_001278518.1 | NP_001265447.1 | 7 | 927 | ||
NM_001278519.1 | NP_001265448.1 | 5 | 737 | ||
NM_002393.4 | NP_002384.2 | 11 | 1406 | ||
XM_017001311.1 | XP_016856800.1 | 11 | 1405 | ||
XM_017001312.1 | XP_016856801.1 | 11 | 1390 | ||
XM_017001313.1 | XP_016856802.1 | 10 | 1255 | ||
GenBank mRNA | AB209383.1 | - | 2 | 2970 | |
AF007111.1 | - | 11 | 1356 | ||
AK223228.1 | - | 11 | 1369 | ||
AK310607.1 | - | 11 | 1382 | ||
AY923176.1 | - | 3 | 316 | ||
AY949028.1 | - | 9 | 1172 | ||
BC067299.1 | - | 11 | 1396 | 30390159 | |
BC105106.1 | - | 10 | 1149 | 8144109 | |
BC143431.1 | - | 10 | 1231 | ||
BC143432.1 | - | 11 | 1328 | ||
BP334671.1 | - | 1 | 126 | ||
BX640923.1 | - | 11 | 1351 | ||
CR749470.1 | - | 10 | 1333 |