Gene Transcripts

Gene Symbol
PMM2
Entrez Gene ID
5373
Gene Name
phosphomannomutase 2
Gene Aliases
CDG1, CDG1a, CDGS, PMI, PMI1, PMM 2
Chromosome Location
Chr. 16: 8797813 - 8849337 on Build GRCh38
UniGene ID
Hs.625732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000303.2 NP_000294.1 5 502
XM_005255374.4 XP_005255431.1 4 425
GenBank mRNA AB209659.1 - 1 3600
AK291537.1 - 5 473
AK296630.1 - 3 279
AK299979.1 - 4 396
AK312532.1 - 5 473
BC008310.1 - 5 444 3611382
DC366756.1 - 5 509
KU178265.1 - 5 432
KU178266.1 - 3 263
U85773.1 - 5 480

View Full Product Details