Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102445.2 | NP_001095915.1 | 6 | 1044 | |
NM_001113380.1 | NP_001106851.1 | 5 | 619 | ||
NM_001113381.1 | NP_001106852.1 | 4 | 488 | ||
NM_005613.5 | NP_005604.1 | 5 | 935 | ||
GenBank mRNA | AB209019.1 | - | 1 | 3011 | |
AF493928.1 | - | 5 | 424 | ||
AK093959.1 | - | 4 | 381 | ||
AK096204.1 | - | 3 | 1549 | ||
AK295540.1 | - | 6 | 779 | ||
AK296240.1 | - | 5 | 463 | ||
AK300832.1 | - | 5 | 423 | ||
AK312250.1 | - | 5 | 547 | ||
BC000737.1 | - | 5 | 509 | 3510260 | |
BC051869.1 | - | 5 | 521 | 5786695 | |
BI547160.1 | - | 5 | 649 | ||
BT007025.1 | - | 5 | 424 | ||
DQ346662.1 | - | 5 | 548 | ||
DQ346663.1 | - | 5 | 763 | ||
DQ346664.1 | - | 6 | 875 | ||
EF054877.1 | - | 5 | 619 | ||
U27768.1 | - | 5 | 521 |