Gene Transcripts

Gene Symbol
MED13
Entrez Gene ID
9969
Gene Name
mediator complex subunit 13
Gene Aliases
ARC250, DRIP250, HSPC221, THRAP1, TRAP240
Chromosome Location
Chr. 17: 61942605 - 62065983 on Build GRCh38
UniGene ID
Hs.282678
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005121.2 NP_005112.2 12 2415
XM_011525551.2 XP_011523853.1 12 3116
XM_011525552.2 XP_011523854.1 12 3116
XM_011525553.2 XP_011523855.1 9 1830
GenBank mRNA AB011165.2 - 9 1833
AF117754.1 - 13 2415
BC140891.1 - 12 2389 9021759

View Full Product Details