Gene Transcripts
- Gene Symbol
- SLC35D2
- Entrez Gene ID
- 11046
- Gene Name
- solute carrier family 35 member D2
- Gene Aliases
- HFRC1, SQV7L, UGTrel8, hfrc
- Chromosome Location
- Chr. 9: 96313437 - 96383710 on Build GRCh38
- UniGene ID
- Hs.494556
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286990.1 |
NP_001273919.1
|
1, 2 |
236 |
|
NM_007001.2 |
NP_008932.2
|
1, 2 |
236 |
|
NR_104627.1 |
|
1, 2 |
236 |
|
XM_005251673.1 |
XP_005251730.1
|
1, 2 |
211 |
|
XM_005251674.1 |
XP_005251731.1
|
1, 2 |
211 |
|
XM_005251675.2 |
XP_005251732.1
|
1, 2 |
221 |
|
XM_005251676.1 |
XP_005251733.1
|
1, 2 |
211 |
|
XM_006716939.2 |
XP_006717002.1
|
1, 2 |
221 |
|
XM_011518164.1 |
XP_011516466.1
|
1, 2 |
221 |
|
GenBank mRNA |
AB106537.1 |
- |
1, 2 |
169 |
|
AB122077.1 |
- |
1, 2 |
181 |
|
AK307890.1 |
- |
1, 2 |
215 |
|
BC100278.1 |
- |
1, 2 |
227 |
6056863 |
BC113579.1 |
- |
2, 3 |
189 |
8322631 |
BX361048.2 |
- |
1, 2 |
235 |
|
View Full Product Details