Gene Transcripts

Gene Symbol
MTO1
Entrez Gene ID
25821
Gene Name
mitochondrial tRNA translation optimization 1
Gene Aliases
CGI-02, COXPD10
Chromosome Location
Chr. 6: 73461727 - 73501456 on Build GRCh38
UniGene ID
Hs.347614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123226.1 NP_001116698.1 13 2209
NM_012123.3 NP_036255.2 12 2089
NM_133645.2 NP_598400.1 13 2164
XM_006715444.2 XP_006715507.1 12 1986
XM_006715445.2 XP_006715508.1 12 1791
XM_006715446.3 XP_006715509.1 11 1928
XM_011535723.1 XP_011534025.1 13 2178
XM_011535724.1 XP_011534026.1 13 2046
XM_011535725.2 XP_011534027.1 13 1914
XM_017010697.1 XP_016866186.1 11 1825
XM_017010698.1 XP_016866187.1 12 2017
XM_017010699.1 XP_016866188.1 11 1511
XM_017010700.1 XP_016866189.1 11 1783
GenBank mRNA AF132937.1 - 12 2011
AF319422.2 - 12 2055
AF469110.1 - 11 1764
AF469111.1 - 11 2145
AJ420530.1 - 11 1998
AK026073.1 - 8 1073
AK074625.1 - 12 2068
AK225828.1 - 13 2193
AL833823.1 - 12 1933
AY078985.1 - 13 2130
AY078986.1 - 13 2130
BC005808.2 - 13 1849
BC011051.1 - 12 2030 4339769

View Full Product Details