Gene Transcripts

Gene Symbol
UNC50
Entrez Gene ID
25972
Gene Name
unc-50 inner nuclear membrane RNA binding protein
Gene Aliases
GMH1, HSD23, UNCL, URP
Chromosome Location
Chr. 2: 98608569 - 98618515 on Build GRCh38
UniGene ID
Hs.13370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014044.5 NP_054763.2 3, 4 550
XM_005263913.4 XP_005263970.1 3, 4 646
XM_006712403.3 XP_006712466.1 3, 4 646
XM_006712404.2 XP_006712467.1 3, 4 560
GenBank mRNA AF077038.1 - 3, 4 502
AK222599.1 - 3, 4 516
AL080115.1 - 3, 4 554
AY017215.1 - 3, 4 518
AY251534.1 - 2, 3 411
BC017019.1 - 3, 4 683 3888349
BC025992.2 - 2, 3 1578 4830706
BC039078.1 - 2, 3 1554 5166015
BC091481.1 - 3, 4 540 30337350
CR533464.1 - 2, 3 398
KU178586.1 - 2, 3 398

View Full Product Details