Gene Transcripts
- Gene Symbol
- LARP7
- Entrez Gene ID
- 51574
- Gene Name
- La ribonucleoprotein domain family member 7
- Gene Aliases
- ALAZS, HDCMA18P, PIP7S
- Chromosome Location
- Chr. 4: 112636964 - 112657592 on Build GRCh38
- UniGene ID
- Hs.713663
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267039.1 |
NP_001253968.1
|
12, 13 |
1912 |
|
NM_015454.2 |
NP_056269.1
|
10, 11 |
1576 |
|
NM_016648.3 |
NP_057732.2
|
10, 11 |
1704 |
|
NR_049768.1 |
|
10, 11 |
1601 |
|
GenBank mRNA |
AF068284.1 |
- |
9, 10 |
853 |
|
AK000274.1 |
- |
10, 11 |
1514 |
|
AK225900.1 |
- |
7, 8 |
1073 |
|
AL049996.1 |
- |
4, 5 |
551 |
|
BC066945.1 |
- |
12, 13 |
1750 |
4837780 |
BC107709.1 |
- |
4, 5 |
545 |
|
BX647100.1 |
- |
10, 11 |
1438 |
|
CA314598.1 |
- |
3 |
596 |
|
EU667388.1 |
- |
10, 11 |
1447 |
|
View Full Product Details