Gene Transcripts

Gene Symbol
LIPT1
Entrez Gene ID
51601
Gene Name
lipoyltransferase 1
Gene Aliases
LIPT1D
Chromosome Location
Chr. 2: 99154955 - 99163157 on Build GRCh38
UniGene ID
Hs.719172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204830.1 NP_001191759.1 3 1091
NM_015929.3 NP_057013.1 3 893
NM_145197.2 NP_660198.1 4 1021
NM_145198.2 NP_660199.1 4 1009
NM_145199.2 NP_660200.1 2 767
NR_037935.1 4 1154
NR_037936.1 3 830
GenBank mRNA AB017566.1 - 3 893
BC007001.1 - 3 813 3961227
BC009772.1 - 3 834 4091817
BC070145.1 - 3 871 4671058

View Full Product Details