Gene Transcripts

Gene Symbol
RC3H2
Entrez Gene ID
54542
Gene Name
ring finger and CCCH-type domains 2
Gene Aliases
MNAB, RNF164
Chromosome Location
Chr. 9: 122849453 - 122905283 on Build GRCh38
UniGene ID
Hs.533499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100588.1 NP_001094058.1 12 2237
NM_018835.2 NP_061323.2 12 2237
GenBank mRNA AF255303.1 - 11 2597
AK000308.1 - 10 1671
AK027042.1 - 3 474
AK226010.1 - 10 1671
AL833177.1 - 13 2094
AL834431.1 - 6 988

View Full Product Details