Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159596.1 | NP_001153068.1 | 2 | 545 | |
NM_017693.3 | NP_060163.2 | 4 | 1229 | ||
GenBank mRNA | AF411385.1 | - | 4 | 1229 | |
AK000166.1 | - | 2 | 441 | ||
AK225229.1 | - | 2 | 441 | ||
AK290658.1 | - | 4 | 1098 | ||
AK304391.1 | - | 3 | 983 | ||
BC020663.1 | - | 2 | 373 | ||
BC039587.1 | - | 2 | 112 | ||
BC051813.2 | - | 2 | 354 | ||
BC075084.2 | - | 2 | 577 | 30915598 | |
BC075085.2 | - | 2 | 577 | 30915386 | |
BC112339.1 | - | 2 | 300 | 40053977 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204425.1 | NP_001191354.1 | 2 | 672 | |
GenBank mRNA | AB209312.1 | - | 2 | 464 | |
LM994822.1 | - | 2 | 550 |