Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159596.1 | NP_001153068.1 | 9 | 1364 | |
NM_017693.3 | NP_060163.2 | 11 | 2027 | ||
GenBank mRNA | AF339783.1 | - | 6 | 618 | |
AF339793.1 | - | 7 | 1416 | ||
AF411385.1 | - | 11 | 2027 | ||
AK000166.1 | - | 9 | 1239 | ||
AK126285.1 | - | 1 | 1040 | ||
AK225229.1 | - | 9 | 1239 | ||
AK290658.1 | - | 11 | 1896 | ||
AK304391.1 | - | 9 | 1648 | ||
BC020663.1 | - | 9 | 1171 | ||
BC051813.2 | - | 9 | 1152 | ||
BC075084.2 | - | 9 | 1375 | 30915598 | |
BC075085.2 | - | 9 | 1375 | 30915386 | |
BC112339.1 | - | 9 | 1119 | 40053977 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001204425.1 | NP_001191354.1 | 9 | 1470 | |
GenBank mRNA | AB209312.1 | - | 9 | 1262 | |
LM994822.1 | - | 9 | 1348 |