Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001310332.1 | NP_001297261.1 | 2 | 129 | |
NM_017999.4 | NP_060469.4 | 2 | 537 | ||
GenBank mRNA | AB265810.1 | - | 2 | 288 | |
AK225832.1 | - | 2 | 487 | ||
AK291247.1 | - | 2 | 489 | ||
AY256461.1 | - | 2 | 117 | ||
BC012077.1 | - | 2 | 457 | 4653017 | |
DA462121.1 | - | 2 | 129 |