Gene Transcripts
- Gene Symbol
- RNLS
- Entrez Gene ID
- 55328
- Gene Name
- renalase, FAD dependent amine oxidase
- Gene Aliases
- C10orf59, RENALASE
- Chromosome Location
- Chr. 10: 88176458 - 88583860 on Build GRCh38
- UniGene ID
- Hs.149849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031709.2 |
NP_001026879.2
|
5, 6 |
835 |
|
NM_018363.3 |
NP_060833.1
|
5, 6 |
835 |
|
XM_005269946.1 |
XP_005270003.1
|
5, 6 |
761 |
|
XM_005269948.2 |
XP_005270005.1
|
3, 4 |
1121 |
|
XM_005269950.3 |
XP_005270007.1
|
3, 4 |
1121 |
|
XM_011539924.2 |
XP_011538226.1
|
5, 6 |
761 |
|
XM_011539927.2 |
XP_011538229.1
|
5, 6 |
761 |
|
XM_017016380.1 |
XP_016871869.1
|
5, 6 |
761 |
|
XM_017016381.1 |
XP_016871870.1
|
5, 6 |
761 |
|
XM_017016382.1 |
XP_016871871.1
|
3, 4 |
1121 |
|
XM_017016385.1 |
XP_016871874.1
|
2, 3 |
1715 |
|
GenBank mRNA |
AK002080.1 |
- |
5, 6 |
762 |
|
AK296262.1 |
- |
3, 4 |
519 |
|
BC005364.1 |
- |
5, 6 |
723 |
3828014 |
BX648154.1 |
- |
2, 3 |
244 |
|
View Full Product Details