Gene Transcripts

Gene Symbol
NXF3
Entrez Gene ID
56000
Gene Name
nuclear RNA export factor 3
Gene Aliases
Chromosome Location
Chr. X: 103075821 - 103093094 on Build GRCh38
UniGene ID
Hs.60386
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022052.1 NP_071335.1 2 139
GenBank mRNA AF346619.1 - 2 139
AJ277527.1 - 2 139
AK302447.1 - 2 140
BC031616.1 - 2 132 5170537

View Full Product Details