Gene Transcripts

Gene Symbol
NIPAL3
Entrez Gene ID
57185
Gene Name
NIPA like domain containing 3
Gene Aliases
DJ462O23.2, NPAL3
Chromosome Location
Chr. 1: 24413360 - 24472983 on Build GRCh38
UniGene ID
Hs.523442
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322854.1 NP_001309783.1 11 1534
NM_001322855.1 NP_001309784.1 12 1676
NM_001322856.1 NP_001309785.1 12 1633
NM_001322857.1 NP_001309786.1 11 1454
NM_001322858.1 NP_001309787.1 12 1551
NM_001322859.1 NP_001309788.1 9 1160
NM_001322860.1 NP_001309789.1 8 1100
NM_001322861.1 NP_001309790.1 8 810
NM_001322862.1 NP_001309791.1 12 1511
NM_001322863.1 NP_001309792.1 11 1483
NM_001322864.1 NP_001309793.1 10 1051
NM_001322866.1 NP_001309795.1 11 1374
NM_001322868.1 NP_001309797.1 9 1248
NM_020448.4 NP_065181.1 11 1396
NR_136506.1 10 1304
NR_136507.1 5 811
NR_136508.1 10 1265
XM_011541808.2 XP_011540110.1 10 1311
XM_017001868.1 XP_016857357.1 10 1046
XM_017001869.1 XP_016857358.1 11 1381
XM_017001870.1 XP_016857359.1 12 1646
GenBank mRNA AK312433.1 - 11 1350
BC063583.1 - 11 1326 4386115
BX538110.1 - 5 424
BX640883.1 - 12 1458
U90918.1 - 11 1287

View Full Product Details