Gene Transcripts

Gene Symbol
NLRC4
Entrez Gene ID
58484
Gene Name
NLR family CARD domain containing 4
Gene Aliases
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF
Chromosome Location
Chr. 2: 32224449 - 32265743 on Build GRCh38
UniGene ID
Hs.574741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199138.1 NP_001186067.1 4 1929
NM_001199139.1 NP_001186068.1 4 1812
NM_021209.4 NP_067032.3 4 1943
XM_017004619.1 XP_016860108.1 4 1929
GenBank mRNA AF376061.1 - 5 2157
AK095467.1 - 4 1929
AK292673.1 - 4 1914
AK314762.1 - 4 1914
AY027787.1 - 4 1944
AY032589.1 - 3 1703
AY035391.1 - 4 1812
AY358152.1 - 4 1891
BC031555.1 - 4 1899 5179909

View Full Product Details