Gene Transcripts
- Gene Symbol
- TMEM43
- Entrez Gene ID
- 79188
- Gene Name
- transmembrane protein 43
- Gene Aliases
- ARVC5, ARVD5, EDMD7, LUMA
- Chromosome Location
- Chr. 3: 14124940 - 14143680 on Build GRCh38
- UniGene ID
- Hs.517817
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024334.2 |
NP_077310.1
|
11, 12 |
1251 |
|
XM_017007176.1 |
XP_016862665.1
|
12, 13 |
1373 |
|
GenBank mRNA |
AK027466.1 |
- |
5, 6 |
481 |
|
AK027827.1 |
- |
6, 7 |
534 |
|
AK027877.1 |
- |
11, 12 |
1115 |
|
AK074073.1 |
- |
9, 10 |
3682 |
|
AK075010.1 |
- |
11, 12 |
1102 |
|
AK225589.1 |
- |
11, 12 |
1174 |
|
AL136916.1 |
- |
11, 12 |
1145 |
|
AY358625.1 |
- |
11, 12 |
1014 |
|
BC003125.1 |
- |
11, 12 |
1146 |
3503636 |
BC008054.2 |
- |
9, 10 |
741 |
|
BC011719.2 |
- |
11, 12 |
1146 |
4079766 |
KU178769.1 |
- |
10, 11 |
997 |
|
KU178770.1 |
- |
9, 10 |
1571 |
|
View Full Product Details