Gene Transcripts
- Gene Symbol
- KAT8
- Entrez Gene ID
- 84148
- Gene Name
- lysine acetyltransferase 8
- Gene Aliases
- MOF, MYST1, ZC2HC8, hMOF
- Chromosome Location
- Chr. 16: 31117664 - 31131393 on Build GRCh38
- UniGene ID
- Hs.533803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032188.2 |
NP_115564.2
|
1, 2 |
226 |
|
NM_182958.2 |
NP_892003.2
|
1, 2 |
226 |
|
GenBank mRNA |
AF217501.1 |
- |
1, 2 |
192 |
|
AF260665.2 |
- |
1, 2 |
124 |
|
AK021872.1 |
- |
1, 2 |
223 |
|
AK024102.1 |
- |
1, 2 |
224 |
|
AK225660.1 |
- |
1, 2 |
226 |
|
AK291106.1 |
- |
1, 2 |
223 |
|
BC037773.1 |
- |
1, 2 |
223 |
5278026 |
BP342476.1 |
- |
1, 2 |
226 |
|
View Full Product Details