Gene Transcripts

Gene Symbol
MFSD7
Entrez Gene ID
84179
Gene Name
major facilitator superfamily domain containing 7
Gene Aliases
LP2561
Chromosome Location
Chr. 4: 676205 - 689468 on Build GRCh38
UniGene ID
Hs.567612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294341.1 NP_001281270.1 7 1254
NM_001294342.1 NP_001281271.1 5 897
NM_032219.3 NP_115595.2 7 1251
XM_005272297.1 XP_005272354.1 5 987
XM_005272299.4 XP_005272356.1 7 1281
XM_006713918.1 XP_006713981.1 7 908
XM_006713919.1 XP_006713982.1 5 990
XM_011513568.1 XP_011511870.1 7 2058
XM_011513571.1 XP_011511873.1 8 1078
XM_011513572.1 XP_011511874.1 5 1074
XM_011513573.2 XP_011511875.1 7 1281
XM_011513574.1 XP_011511876.1 6 1047
XM_011513575.2 XP_011511877.1 6 1826
XM_017008700.1 XP_016864189.1 6 1161
GenBank mRNA AK025922.1 - 5 729
AK292010.1 - 7 1149
AY203936.1 - 3 600
AY358401.1 - 7 956
BC030246.1 - 7 994 5216645

View Full Product Details