Gene Transcripts

Gene Symbol
OR4D2
Entrez Gene ID
124538
Gene Name
olfactory receptor family 4 subfamily D member 2
Gene Aliases
BC2009, OR17-24
Chromosome Location
Chr. 17: 58169656 - 58170579 on Build GRCh38
UniGene ID
Hs.446879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004707.3 NP_001004707.1 1 21
GenBank mRNA AF303373.1 - 1 276

View Full Product Details