Gene Transcripts

Gene Symbol
OR6N1
Entrez Gene ID
128372
Gene Name
olfactory receptor family 6 subfamily N member 1
Gene Aliases
OR1-22
Chromosome Location
Chr. 1: 158747733 - 158840979 on Build GRCh38
UniGene ID
Hs.553592
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005185.1 NP_001005185.1 1 422
XM_017000325.1 XP_016855814.1 3 1280
XM_017000326.1 XP_016855815.1 4 1861
XM_017000327.1 XP_016855816.1 3 654
GenBank mRNA BC146847.1 - 1 442

View Full Product Details