Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286808.1 | NP_001273737.1 | 6 | 1093 | |
NM_130809.4 | NP_570721.1 | 6 | 1093 | ||
XM_006714529.3 | XP_006714592.1 | 6 | 1301 | ||
GenBank mRNA | AJ427339.1 | - | 6 | 1046 | |
AJ441110.3 | - | 6 | 1046 | ||
AJ496322.1 | - | 6 | 1046 | ||
AJ515158.1 | - | 6 | 1046 | ||
AJ515428.1 | - | 6 | 1046 | ||
AJ515429.1 | - | 6 | 1046 | ||
AK057437.1 | - | 6 | 1030 | ||
AK090486.1 | - | 5 | 918 | ||
AL831963.1 | - | 6 | 1055 | ||
AL832056.1 | - | 6 | 1056 | ||
AL832600.2 | - | 6 | 1052 | ||
BC017066.1 | - | 6 | 1030 | 3913311 | |
BX099255.1 | - | 3 | 289 |