Gene Transcripts

Gene Symbol
OR1L8
Entrez Gene ID
138881
Gene Name
olfactory receptor family 1 subfamily L member 8
Gene Aliases
OR9-24
Chromosome Location
Chr. 9: 122546009 - 122576960 on Build GRCh38
UniGene ID
Hs.632695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004454.1 NP_001004454.1 1 573
XM_017014284.1 XP_016869773.1 3 1109
XM_017014285.1 XP_016869774.1 3 1109
XM_017014286.1 XP_016869775.1 2 980

View Full Product Details