Gene Transcripts

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
Chromosome Location
Chr. 9: 36336398 - 36487384 on Build GRCh38
UniGene ID
Hs.333503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022781.4 NP_073618.3 3 453
NM_194328.2 NP_919309.1 3 620
NM_194329.2 NP_919310.1 2 303
NM_194330.2 NP_919311.1 3 351
NM_194332.2 NP_919313.1 2 470
XM_005251366.3 XP_005251423.1 3 457
XM_005251367.3 XP_005251424.1 3 355
XM_005251368.3 XP_005251425.1 3 469
XM_006716721.3 XP_006716784.1 2 317
XM_011517712.2 XP_011516014.1 4 393
XM_011517713.2 XP_011516015.1 3 457
XM_017014295.1 XP_016869784.1 3 484
GenBank mRNA AF394047.1 - 3 575
AK097653.1 - 3 319
AL136817.1 - 3 371
BC033786.1 - 2 303 5166325
BG722531.1 - 2 475
BI520161.1 - 3 349
DA189510.1 - 2 421
DA483270.1 - 2 201

View Full Product Details