Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172250.2 | NP_758454.1 | 2 | 222 | |
XM_011531684.2 | XP_011529986.1 | 2 | 300 | ||
XM_011531685.2 | XP_011529987.1 | 2 | 343 | ||
XM_011531686.2 | XP_011529988.1 | 1 | 202 | ||
GenBank mRNA | AK126662.1 | - | 2 | 222 | |
BC101178.2 | - | 1 | 184 | 40017262 | |
BC101179.2 | - | 1 | 184 | 40017263 | |
BC101180.2 | - | 1 | 184 | ||
BC101181.2 | - | 1 | 184 | 40017265 |