Gene Transcripts
- Gene Symbol
- PADI6
- Entrez Gene ID
- 353238
- Gene Name
- peptidyl arginine deiminase 6
- Gene Aliases
- hPADVI
- Chromosome Location
- Chr. 1: 17372196 - 17401699 on Build GRCh38
- UniGene ID
- Hs.531598
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207421.4 |
NP_997304.3
|
13 |
1602 |
|
GenBank mRNA |
AY422079.1 |
- |
13 |
1552 |
|
View Full Product Details