Gene Transcripts

Gene Symbol
MYL6
Entrez Gene ID
4637
Gene Name
myosin light chain 6
Gene Aliases
ESMLC, LC17, LC17-GI, LC17-NM, LC17A, LC17B, MLC-3, MLC1SM, MLC3NM, MLC3SM
Chromosome Location
Chr. 12: 56158261 - 56161582 on Build GRCh38
UniGene ID
Hs.632717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021019.4 NP_066299.2 3, 4 314
NM_079423.3 NP_524147.2 3, 4 314
GenBank mRNA AB046613.1 - 3, 4 213
AK301198.1 - 2, 3 492
BC006781.2 - 1, 2 144
BC017455.1 - 3, 4 201 3538373
BC071661.1 - 3, 4 197 5561575
BC093066.1 - 3, 4 210 30722094
BG283793.1 - 3, 4 314
CA427580.1 - 4, 5 510
CN401857.1 - 3, 4 198
CR456839.1 - 1, 2 68
CR457014.1 - 2, 3 173
M22918.1 - 3, 4 204
M22920.1 - 2, 3 182
M31212.1 - 3, 4 213
U02629.1 - 3, 4 211

View Full Product Details